Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse HMGB3 Alexa Fluor Plus 680-conjugated Antibody, R&D Systems™
SDP

Catalog No. RD10164204
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10164204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10164204 Supplier R&D Systems Supplier No. FAB55071AFP680100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

HMGB3/HMG4 Monoclonal antibody specifically detects HMGB3/HMG4 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry

Specifications

Antigen HMGB3/HMG4
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 546519
Conjugate Alexa Fluor Plus 680
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2A, HMG2AHMG-4, HMG4, HMG4MGC90319, non-histone chromosomal protein
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3149
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied
Product Type Antibody
Form Liquid
Isotype IgG1
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.