Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse IGF-I R/IGF1R Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB391100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB391100 100 μg
MAB391SP 25 μg
MAB391500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB391100 Supplier R&D Systems Supplier No. MAB391100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 17 publications

IGF-I R/IGF1R Monoclonal specifically detects IGF-I R/IGF1R in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen IGF-I R
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, ELISA
Classification Monoclonal
Clone 33255
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Neutralization, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P08069
Gene Alias CD221, CD221 antigen, EC 2.7.10, EC 2.7.10.1, IGF1R, IGF-1R, IGF-I receptor, IGFIR, IGF-IR, IGFR, insulin-like growth factor 1 receptor, Insulin-like growth factor I receptor, JTK13, MGC142170, MGC142172, MGC18216, soluble IGF1R variant 1, soluble IGF1R variant 2
Gene Symbols IGF1R
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IGF-I R Glu31-Asn932 Accession # P08069
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3480
Test Specificity Detects human IGF-I R in sandwich ELISAs and Western blots. Detects mouse IGF-I R in Immunohistochemistry. In sandwich immunoassays, less than 0.15% cross-reactivity or interference was observed with recombinant human (rh) IGF-I, rhIGF-II, rhIL-3 R alpha , rhIL-9 R, and rhTGF-beta RII.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.