Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Neurexin 3/NRXN3 Antibody, R&D Systems™

Catalog No. AF5269
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5269 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5269 Supplier R&D Systems Supplier No. AF5269
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Neurexin 3/NRXN3 Polyclonal specifically detects Neurexin 3/NRXN3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Neurexin 3/NRXN3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. NP_620426
Gene Alias C14orf60, chromosome 14 open reading frame 60, KIAA0743MGC176711, neurexin 3, neurexin III, Neurexin III-alpha, Neurexin III-beta, neurexin-3-beta, NRXN3
Gene Symbols NRXN3
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Neurexin 3/NRXN3 Ser35-Thr357 Accession # NP_620426
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9369
Test Specificity Detects human and mouse Neurexin 3/NRXN3 in Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Neurexin 1 beta is observed and less than 5% cross-reactivity recombinant rat (rr) Neurexin 1 alpha and rrNeurexin 2 beta is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.