Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Primate BMP-9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-4711995
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3209SP 25 μg
MAB3209100 100 μg
MAB3209500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB3209SP Supplier R&D Systems Supplier No. MAB3209SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 11 publications

BMP-9 Monoclonal specifically detects BMP-9 in Human, Mouse, Primate samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen BMP-9
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 360107
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.01-0.04 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UK05
Gene Alias BMP9, BMP9BMP-9Bone morphogenetic protein 9, GDF2, GDF-2, growth differentiation factor 2, growth/differentiation factor 2
Gene Symbols GDF2
Host Species Mouse
Immunogen E. coli-derived recombinant human BMP-9 Ser320-Arg429 Accession # Q9UK05
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2658
Test Specificity Detects human and primate BMP-9 in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human BMP-1, -1.1, -2, -3, -3b, -4, -5, -6, -7, -8, or -10 is observed. In direct ELISAs, approximately 80% cross-reactivity with recombinant mouse (rm) BMP-9 and 10% cross-reactivity with rmBMP-10 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Primate
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.