Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat Activin A Protein
GSA_VASDP

Catalog No. 338AC500CF Shop All R&D Systems Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
33-8AC-500CF Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
33-8AC-500 Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-8AC-500CF Supplier R&D Systems™ Supplier No. 338AC500/CF
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

CHO derived

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL.

Order Info

Ships at ambient temperature.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accession Number P08476
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 3624
Molecular Weight (g/mol) M.W. (Observed): 14 kDa, reducing conditions 24 kDa, non-reducing conditions; M.W. (theoretical): 13 kDa (monomer)
Quantity 500 μg
Source Chinese Hamster Ovary cell line, CHO-derived Activin A protein Gly311-Ser426
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol INHBA
Biological Activity 0.2 to 1.2ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human, Mouse, Rat
Recombinant Recombinant
Reconstitution Reconstitute at 100-500 μg/mL in sterile 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining.
Protein Activin A
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.