Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Casein Kinase 1 delta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4568
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4568 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4568 Supplier R&D Systems Supplier No. AF4568
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Casein Kinase 1 delta Polyclonal specifically detects Casein Kinase 1 delta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Casein Kinase 1 delta
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P48730
Gene Alias casein kinase 1, delta, casein kinase I isoform delta, CKId, CKI-delta, CSNK1D, EC 2.7.11, EC 2.7.11.1, HCKID
Gene Symbols CSNK1D
Host Species Sheep
Immunogen E. coli-derived recombinant human CK1 delta Arg316-Arg415 Accession # P48730
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1453
Test Specificity Detects endogenous human, mouse and rat CK1 delta in Western blots. In Western blots, this antibody does not cross-react with recombinant human CK1 alpha or rhCK1 gamma ; reactivity with CK1 epsilon is unknown.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.