Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat GDF-11/GDF-8 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB788100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB788100 100 μg
MAB788SP 25 μg
MAB788500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB788100 Supplier R&D Systems Supplier No. MAB788100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 8 publications

GDF-11/GDF-8 Monoclonal specifically detects GDF-11/GDF-8 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GDF-11/GDF-8
Applications Western Blot
Classification Monoclonal
Clone 84214
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O08689
Gene Symbols GDF11
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse GDF-8 Asn25-Ser376 Accession # O08689
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10220
Test Specificity Detects human, mouse, and rat GDF-8 in direct ELISAs and Western blots. In Western blots, this antibody shows approximately 30% cross-reactivity with recombinant human/mouse/rat GDF-11. In direct ELISAs, this antibody shows less than 1% cross-reactivity with recombinant mouse (rm) GDF-1, rmGDF-3, rmGDF-5, rmGDF-6, and rmGDF-9.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.