Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat HSP60 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1800
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1800 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1800 Supplier R&D Systems Supplier No. MAB1800
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

HSP60 Monoclonal specifically detects HSP60 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.

Specifications

Antigen HSP60
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 264233
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10809
Gene Alias 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HSPD1, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant), SPG13
Gene Symbols HSPD1
Host Species Mouse
Immunogen E. coli-derived recombinant human HSP60 Met1-Phe573 Accession # P10809
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3329
Test Specificity Detects human, mouse and rat HSP60.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.