Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat IGSF8/CD316 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7237024
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB31171SP 25 μg
MAB31171100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB31171SP Supplier R&D Systems Supplier No. MAB31171SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IGSF8/CD316 Monoclonal antibody specifically detects IGSF8/CD316 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen IGSF8/CD316
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2587G
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q969P0-1
Gene Alias CD316, CD316 antigen, CD81P3, CD81P3LIR-D1, EWI2, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRL, PGRLimmunoglobulin superfamily member 8
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-derived human IGSF8/CD316, Arg28-Thr579, Accession # Q969P0-1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93185
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.