Learn More
Human/Mouse/Rat IMPA1/IMP1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IMPA1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 984603 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1μg/mL |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. |
| Gene Accession No. | P29218 |
| Gene Alias | EC 3.1.3.25, IMP, IMP 1, IMP1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1, MRT59, Myo-Inositol Monophosphatase 1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.