Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Importin beta/KPNB1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8209SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8209SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8209SP Supplier R&D Systems Supplier No. MAB8209SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Importin beta/KPNB1 Monoclonal specifically detects Importin beta/KPNB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Importin beta/KPNB1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 845208
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q14974
Gene Alias IMB1, Impnb, importin 1, importin 90, importin beta-1 subunit, importin subunit beta-1, importin-90, IPO1, IPOB, karyopherin (importin) beta 1, Karyopherin subunit beta-1, KPNB1, MGC2155, MGC2156, MGC2157, NTF97, NTF97Pore targeting complex 97 kDa subunit, Nuclear factor p97, PTAC97
Gene Symbols KPNB1
Host Species Mouse
Immunogen E. coli-derived recombinant human Importin beta/KPNB1 Met1-Gly155 Accession # Q14974
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3837
Test Specificity Detects human Importin beta/KPNB1 in ELISA and Western Blot. It detects mouse and rat Importin beta/KPNB1 in Western Blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.