Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human/Mouse/Rat Irisin Recombinant Protein, PeproTech®

Catalog No. 10065250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10065250UG 250 μg
10065100UG 100 μg
100-65-10UG 10 μg
100-65-1MG 1 mg
100-65-2UG 2 μg
10065500UG 500 μg
100-65-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 10065250UG Supplier Gibco™ Supplier No. 10065250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human/Murine/Rat Irisin, appears to form a non-disulfide linked oligomeric structure in solution, has a calculated theoretical molecular weight of 25.2 kDa, but migrates at an apparent molecular weight of 28 kDa by SDS-PAGE analysis under reducing conditions due to glycosylation. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8K3V5, Q8K4Z2, Q8NAU1
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 252995, 260327, 384061
Molecular Weight (g/mol) 25.2 kDa
Name Human/Mouse/Rat Irisin
Quantity 250 μg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 1500001L03Rik; AI836596; C87088; fibronectin type III domain containing 5; fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Fndc5; Frcp2; Irisin; PeP; Peroxisomal protein; Pxp
Common Name FNDC5
Gene Symbol FNDC5
Biological Activity Determined by its ability to promote cell survival and growth of rat H9c2 cardio-myoblasts under serum starvation conditions.
Conjugate Unconjugated
Recombinant Recombinant
Sequence DSPSAPVNVT VRHLKANSAV VSWDVLEDEV VIGFAISQQK KDVRMLRFIQ EVNTTTRSCA LWDLEEDTEY IVHVQAISIQ GQSPASEPVL FKTPREAEKM ASKNKDEVTM KE
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.