Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat MKP-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3576
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3576 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3576 Supplier R&D Systems Supplier No. MAB3576
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MKP-3/DUSP6 Monoclonal specifically detects MKP-3/DUSP6 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MKP-3/DUSP6
Applications Western Blot
Classification Monoclonal
Clone 363246
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q16828
Gene Alias dual specificity phosphatase 6, Dual specificity protein phosphatase PYST1, DUSP6, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP3, MKP-3, MKP3serine/threonine specific protein phosphatase, PYST1, PYST1dual specificity protein phosphatase 6
Gene Symbols DUSP6
Host Species Mouse
Immunogen E. coli-derived recombinant human MKP-3 Ile2-Thr381 Accession # Q16828
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1848
Test Specificity Detects endogenous human, mouse, and rat MKP-3 in Western blots. In Western blots, this antibody does not cross-react with recombinant human MKP-1 or MKP-2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.