Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human/Mouse/Rat Myostatin Recombinant Protein, PeproTech®

Catalog No. 120001MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
120-00-1MG 1 mg
12000100UG 100 μg
120-00-10UG 10 μg
12000250UG 250 μg
120-00-2UG 2 μg
12000500UG 500 μg
120-00-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 120-00-1MG Supplier Gibco™ Supplier No. 120001MG
Add to Cart
Edge
Add to Cart
Request a Quote
This item is not returnable. View return policy

Recombinant Protein

The amino acid sequence of mature myostatin is extremely conserved across species, and is the same in murine, rat, chicken, turkey, porcine, and human. Myostatin is expressed as the C-terminal part of a precursor polypeptide, which also contains a short N-terminal signal sequence for secretion, and a propeptide of 243 amino acids. After dimerization of this precursor, the covalent bonds between the propeptide and the mature ligand are cleaved by furin-type proteases. However, the resulting two proteins remain associated through non-covalent interactions, and are secreted as a latent complex. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O08689, O14793, O35312
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 17700, 2660, 29152
Molecular Weight (g/mol) 25 kDa
Name Human/Mouse/Rat Myostatin
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Cmpt; compact; Gdf8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; Myostatin
Common Name Myostatin
Gene Symbol Mstn
Biological Activity Determined by its ability to inhibit the proliferation of MPC-11 cells. The expected ED50 for this effect is 17.0-25.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence DFGLDCDEHS TESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGECEFV FLQKYPHTHL VHQANPRGSA GPCCTPTKMS PINMLYFNGK EQIIYGKIPA MVVDRCGCS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.