Learn More
Description
Includes
25μg each of antibodies to Notch-1, CXCR4, Vimentin, SSEA-1, Musashi-1, SOX1, SOX2, and Nestin supplied as 10X stock
Includes eight antibodies to increase confidence in cell identification and characterization
Compatible with detection of SSEA-1 and CXCR4 by flow cytometry, which can be completed in less than two hours
Compatible with detection of all markers by immun
Specifications
Specifications
| Cross Reactivity | Mammals |
| Quantity | 1 Kit |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Product Line | R&D Systems |
| Product Type | Neural Progenitor Markers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
