Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat Phospho-JNK2 (T183/Y185) DuoSet IC ELISA

Catalog No. p-4754638
Change view
Click to view available options
Format:
15 96-well Plates
2 96-well Plates
5 96-well Plates
Gene ID (Entrez):
5601
50658
243911
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Gene ID (Entrez)
DYC22362 2 96-well Plates 243911
DYC22365 5 96-well Plates 5601
DYC2236E 15 96-well Plates 50658
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. DYC22362 Supplier R&D Systems™ Supplier No. DYC22362
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human/Mouse/Rat Phospho-JNK2 (T183/Y185) DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human/Mouse/Rat Phospho-JNK2 (T183/Y185) DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat JNK2.

Specifications

Product Type ELISA Development Kit for Phosphorylated Intracellular Proteins
Quantity 1 Kit (for 2 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 125.00 to 8000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 2 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 243911
Synonym c-Jun kinase 2, c-Jun N-terminal kinase 2, EC 2.7.11, EC 2.7.11.24, JNK2A, JNK2BETA, JNK2JNK2ALPHA, JNK-55, Jun kinase, MAP kinase 9, MAPK 9, MAPK9, mitogen-activated protein kinase 9, p54a, p54aSAPK, PRKM9JNK2B, SAPK, SAPK1 alpha, Stress-activated protein kinase JNK2
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.