Learn More
R&D Systems™ Human/Mouse/Rat/Porcine/Canine TGF-beta 1 Quantikine ELISA
Shop All Novus Biologicals Products

Specifications
Specifications
| Product Line | R&D Systems |
| Specificity | Natural and recombinant TGF-beta 1. This assay also recognizes human TGF-beta 1.2. |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7040 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | CEDLAP, DPD1, latency-associated peptide, TGF beta, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.