Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human/Mouse/Rat RelA/NFkB p65 ELISA Kit
SDP

Catalog No. NBP2296611 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2296611 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2296611 Supplier Novus Biologicals™ Supplier No. NBP2296611KIT
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human, Mouse, Rat RelA/NFkB p65 ELISA Kit (Colorimetric) measures human, mouse, rat RelA/NFkB p65 in cell and tissue lysates.

Specifications

Product Type ELISA Kit
Target Species Human, Mouse, Rat
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 5970
Gene Symbol RELA
Kit Contents Capture Antibody 200ul, Detecting Antibody 200μL, Coating Buffer 2 x 10mL, BSA 2 x 0.5g, AKP-Conjugated Secondary Ab 10μL, pNPP Substrate Buffer 2 x 10mL, Recombinant p65 Standard 2 vials lyophilized (0.42 ug/vial), pNPP 4 x 5 mg, ELISA Plates 2, Manual 1, 10X Hypotonic Lysis Buffer 10mL, 1X Nuclear Extraction Buffer 10mL, 10% Detergent Solution 10mL, 10X PBS 2 x 50mL, 1 M DTT (for Nuclear Extraction from tissue) 100μL, 10 mM DTT (for Nuclear Lysis Buffer) 500μL, 100X Protease Inhibitor Cocktail (PIC) 100μL, 100 mM PMSF 10 ml
Quantity 1 Kit
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators
Storage Requirements Storage is content dependent.
Synonym MGC131774, nf kb elisa kit, nf kb p65, nfkb elisa kit, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit,
Target RelA/NFkB p65
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.