Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human/Mouse/Rat Total ERK2 DuoSet IC ELISA

Catalog No. DYC1230C5
Change view
Click to view available options
Format:
2 96-well Plates
5 96-well Plates
Gene ID (Entrez):
5594
26413
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Gene ID (Entrez)
DYC1230C5 5 96-well Plates 26413
DYC1230C2 2 96-well Plates 5594
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. DYC1230C5 Supplier R&D Systems™ Supplier No. DYC1230C5
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human/Mouse/Rat Total ERK2 DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human/Mouse/Rat Total ERK2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat ERK2.

Specifications

Product Type ELISA Development Kit for Intracellular Proteins
Quantity 1 Kit (for 5 Plates)
Species Human, Mouse, Rat
Conjugate Biotin
Assay Range 78.10 to 5000 pg/mL
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Format 5 96-well Plates
For Use With (Application) ELISA
Gene ID (Entrez) 26413
Synonym EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.