Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Vimentin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2105SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2105SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2105SP Supplier R&D Systems Supplier No. MAB2105SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 9 publications

Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, Knockout Validated.

Specifications

Antigen Vimentin
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, Immunoassay
Classification Monoclonal
Clone 280618
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 0.5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P08670
Gene Alias FLJ36605, VIM, vimentin
Gene Symbols VIM
Host Species Rat
Immunogen E. coli-derived recombinant human Vimentin Ser2-Glu466 Accession # P08670
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7431
Test Specificity Vimentin antibodies are ideal for immunocytochemistry colocalization studies in intermediate filaments. Detects human, mouse and rat Vimentin in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.