Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse RBP4/Retinol-Binding Protein 4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB34761SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB34761SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB34761SP Supplier R&D Systems Supplier No. MAB34761SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

RBP4/Retinol-Binding Protein 4 Monoclonal specifically detects RBP4/Retinol-Binding Protein 4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen RBP4/Retinol-Binding Protein 4
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 423619
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.05-2 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_035385
Gene Alias interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, RetinolBinding Protein 4, retinol-binding protein 4, plasma
Gene Symbols RBP4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4 Met1-Leu201 Accession # NP_035385
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5950
Test Specificity Detects mouse RBP4/Retinol-Binding Protein 4 in direct ELISA and Western Blot. In Western blots, detects human RBP4/Retinol-Binding Protein 4.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.