Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Wnt-2b Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7061995
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3900SP 25 μg
AF3900 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3900SP Supplier R&D Systems Supplier No. AF3900SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Wnt-2b Polyclonal specifically detects Wnt-2b in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Wnt-2b
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O70283
Gene Alias Protein Wnt-13, protein Wnt-2b, wingless-type MMTV integration site family, member 2B, Wnt-13, WNT13XWNT2, Xenopus, homolog of, Wnt2b, XWNT2, XWNT2wingless-type MMTV integration site family, member 13
Gene Symbols WNT2B
Host Species Goat
Immunogen E. coli-derived recombinant mouse Wnt-2b Leu64-Glu104 and Arg251-Ala312 Accession # O70283
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7482
Test Specificity Detects recombinant human and recombinant mouse Wnt-2b in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Wnt-2 is observed and less than 5% cross-reactivity with rhWnt-7a, -9a, recombinant mouse Wnt-1, -3a, -4, -5a, -5b, -8a, -8b, -9b, -10a, -10b, -11, and -16 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.