Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MPHOSPH1 Partial ORF (NP_057279, 1671 a.a. - 1780 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89952813 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-813 10 μg
89-952-814 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-952-813 Supplier Abnova™ Supplier No. H00009585Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Sequence: RSQASIIGVNLATKKKEGTLQKFGDFLQHSPSILQSKAKKIIETMSSSKLSNVEASKENVSQPKRAKRKLYTSEISSPIDISGQVILMDQKMKESDHQIIKRRLRTKTAK

Specifications

Accession Number NP_057279
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9585
Molecular Weight (g/mol) 37.84kDa
Name MPHOSPH1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen RSQASIIGVNLATKKKEGTLQKFGDFLQHSPSILQSKAKKIIETMSSSKLSNVEASKENVSQPKRAKRKLYTSEISSPIDISGQVILMDQKMKESDHQIIKRRLRTKTAK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias DKFZp434B0435/DKFZp434P0810/KRMP1/MPHOSPH1/MPP-1/MPP1
Common Name KIF20B
Gene Symbol KIF20B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.