Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MPP5 Full-length ORF (NP_071919.2, 1 a.a. - 675 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89952841 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-841 25 μg
89-952-840 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-952-841 Supplier Abnova™ Supplier No. H00064398P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like (see DLG1; MIM 601014), ZO1-like (see TJP1; MIM 601009), p55-like (see MPP1; MIM 305360), and LIN2-like (see CASK; MIM 300172), based on their size and the presence of additional domains (Tseng et al., 2001 [PubMed 11311936]). MPP5 is a member of the p55-like MAGUK subfamily.[supplied by OMIM]

Sequence: MTTSHMNGHVTEESDSEVKNVDLASPEEHQKHREMAVDCPGDLGTRMMPIRRSAQLERIRQQQEDMRRRREEEGKKQELDLNSSMRLKKLAQIPPKTGIDNPMFDTEEGIVLESPHYAVKILEIEDLFSSLKHIQHTLVDSQSQEDISLLLQLVQNKDFQNAFKIHNAITVHMNKASPPFPLISNAQDLAQEVQTVLKPVHHKEGQELTALLNTPHIQALLLAHDKVAEQEMQLEPITDERVYESIGQYGGETVKIVRIEKARDIPLGATVRNEMDSVIISRIVKGGAAEKSGLLHEGDEVLEINGIEIRGKDVNEVFDLLSDMHGTLTFVLIPSQQIKPPPAKETVIHVKAHFDYDPSDDPYVPCRELGLSFQKGDILHVISQEDPNWWQAYREGDEDNQPLAGLVPGKSFQQQREAMKQTIEEDKEPEKSGKLWCAKKNKKKRKKVLYNANKNDDYDNEEILTYEEMSLYHQPANRKRPIILIGPQNCGQNELRQRLMNKEKDRFASAVPHTTRSRRDQEVAGRDYHFVSRQAFEADIAAGKFIEHGEFEKNLYGTSIDSVRQVINSGKICLLSLRTQSLKTLRNSDLKPYIIFIAPPSQERLRALLAKEGKNPKPEELREIIEKTREMEQNNGHYFDTAIVNSDLDKAYQELLRLINKLDTEPQWVPSTWLR

Specifications

Accession Number NP_071919.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 64398
Molecular Weight (g/mol) 103.7kDa
Name MPP5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MTTSHMNGHVTEESDSEVKNVDLASPEEHQKHREMAVDCPGDLGTRMMPIRRSAQLERIRQQQEDMRRRREEEGKKQELDLNSSMRLKKLAQIPPKTGIDNPMFDTEEGIVLESPHYAVKILEIEDLFSSLKHIQHTLVDSQSQEDISLLLQLVQNKDFQNAFKIHNAITVHMNKASPPFPLISNAQDLAQEVQTVLKPVHHKEGQELTALLNTPHIQALLLAHDKVAEQEMQLEPITDERVYESIGQYGGETVKIVRIEKARDIPLGATVRNEMDSVIISRIVKGGAAEKSGLLHEGDEVLEINGIEIRGKDVNEVFDLLSDMHGTLTFVLIPSQQIKPPPAKETVIHVKAHFDYDPSDDPYVPCRELGLSFQKGDILHVISQEDPNWWQAYREGDEDNQPLAGLVPGKSFQQQREAMKQTIEEDKEPEKSGKLWCAKKNKKKRKKVLYNANKNDDYDNEEILTYEEMSLYHQPANRKRPIILIGPQNCGQNELRQRLMNKEKDRFASAVPHTTRSRRDQEVAGRDYHFVSRQAFEADIAAGKFIEHGEFEKNLYGTSIDSVRQVINSGKICLLSLRTQSLKTLRNSDLKPYIIFIAPPSQERLRALLAKEGKNPKPEELREIIEKTREMEQNNGHYFDTAIVNSDLDKAYQELLRLINKLDTEPQWVPSTWLR
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ12615/PALS1
Common Name MPP5
Gene Symbol MPP5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.