Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MRPL49 Full-length ORF (NP_004918.1, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89952943 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-943 25 ug
89-952-942 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-952-943 Supplier Abnova™ Supplier No. H00000740P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene and the gene for the HRD1 protein use in their respective 3' UTRs some of the same genomic sequence. Pseudogenes corresponding to this gene are found on chromosomes 5q and 8p. [provided by RefSeq]

Sequence: MAATMFRATLRGWRTGVQRGCGLRLLSQTQGPPDYPRFVESVDEYQFVERLLPATRIPDPPKHEHYPTPSGWQPPRDPPPNLPYFVRRSRMHNIPVYKDITHGNRQMTVIRKVEGDIWALQKDVEDFLSPLLGKTPVTQVNEVTGTLRIKGYFDQELKAWLLEKGF

Specifications

Accession Number NP_004918.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 740
Molecular Weight (g/mol) 45.6kDa
Name MRPL49 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MAATMFRATLRGWRTGVQRGCGLRLLSQTQGPPDYPRFVESVDEYQFVERLLPATRIPDPPKHEHYPTPSGWQPPRDPPPNLPYFVRRSRMHNIPVYKDITHGNRQMTVIRKVEGDIWALQKDVEDFLSPLLGKTPVTQVNEVTGTLRIKGYFDQELKAWLLEKGF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias C11orf4/L49mt/MGC10656/NOF/NOF1
Common Name MRPL49
Gene Symbol MRPL49
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.