Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MS4A2 Partial ORF (NP_000130, 1 a.a. - 59 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953001 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-001 10 ug
89-953-002 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-001 Supplier Abnova™ Supplier No. H00002206Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The allergic response involves the binding of allergen to receptor-bound IgE followed by cell activation and the release of mediators responsible for the manifestations of allergy. The IgE-receptor, a tetramer composed of an alpha, beta, and 2 disulfide-linked gamma chains, is found on the surface of mast cells and basophils. This gene encodes the beta subunit of the high affinity IgE receptor which is a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This family member is localized to 11q12, among a cluster of family members. Alternative splicing results in multiple transcript variants encoding different isoforms

Sequence: MDTESNRRANLALPQEPSSVPAFEVLEISPQEVSSGRLLKSASSPPLHTWLTVLKKEQE

Specifications

Accession Number NP_000130
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2206
Molecular Weight (g/mol) 32.23kDa
Name MS4A2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MDTESNRRANLALPQEPSSVPAFEVLEISPQEVSSGRLLKSASSPPLHTWLTVLKKEQE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias APY/ATOPY/FCER1B/FCERI/IGEL/IGER/IGHER/MS4A1
Common Name MS4A2
Gene Symbol MS4A2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.