Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MS4A6E Full-length ORF (NP_640342.1, 1 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953015 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-015 10 μg
89-953-016 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-015 Supplier Abnova™ Supplier No. H00245802P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. The gene encoding this protein is localized to 11q12.3, among a cluster of family members. [provided by RefSeq]

Sequence: MTSQPISNETIIMLPSNVINFSQAEKPEPTNQGQDSLKKRLQAKVKVIGVHSSLAGSILSALSALVGFILLSVNPAALNPASLQCKLDEKDIPTRLLLSYDYHSPYTMDCHRAKASLAGTLSLMLVSTVLEFCLAVLTAVLQWKQTV

Specifications

Accession Number NP_640342.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 245802
Molecular Weight (g/mol) 42.3kDa
Name MS4A6E (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MTSQPISNETIIMLPSNVINFSQAEKPEPTNQGQDSLKKRLQAKVKVIGVHSSLAGSILSALSALVGFILLSVNPAALNPASLQCKLDEKDIPTRLLLSYDYHSPYTMDCHRAKASLAGTLSLMLVSTVLEFCLAVLTAVLQWKQTV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC126832
Common Name MS4A6E
Gene Symbol MS4A6E
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.