Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MSH6 Partial ORF (NP_000170, 931 a.a. - 1030 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953041 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-041 25 ug
89-953-040 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-041 Supplier Abnova™ Supplier No. H00002956Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a protein similar to the MutS protein. In E. coli, the MutS protein helps in the recognition of mismatched nucleotides, prior to their repair. A highly conserved region of approximately 150 aa, called the Walker-A adenine nucleotide binding motif, exists in MutS homologs. The encoded protein of this gene combines with MSH2 to form a mismatch recognition complex that functions as a bidirectional molecular switch that exchanges ADP and ATP as DNA mismatches are bound and dissociated. Mutations in this gene have been identified in individuals with hereditary nonpolyposis colon cancer (HNPCC) and endometrial cancer. [provided by RefSeq]

Sequence: AGFDSDYDQALADIRENEQSLLEYLEKQRNRIGCRTIVYWGIGRNRYQLEIPENFTTRNLPEEYELKSTKKGCKRYWTKTIEKKLANLINAEERRDVSLK

Specifications

Accession Number NP_000170
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2956
Molecular Weight (g/mol) 36.74kDa
Name MSH6 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen AGFDSDYDQALADIRENEQSLLEYLEKQRNRIGCRTIVYWGIGRNRYQLEIPENFTTRNLPEEYELKSTKKGCKRYWTKTIEKKLANLINAEERRDVSLK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GTBP/HNPCC5/HSAP
Common Name MSH6
Gene Symbol MSH6
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.