Learn More
Abnova™ Human MSL3L1 Full-length ORF (AAH31210.1, 1 a.a. - 521 a.a.) Recombinant Protein with GST-tag at N-terminal
Shop All Abnova Corporation ProductsDescription
This gene encodes a nuclear protein and has similarity to drosophila male-specific lethal-3 gene. The drosophila protein plays a critical role in a dosage-compensation pathway, which equalizes X-linked gene expression in males and females. Thus this encoded protein is thought to play a similar function in chromatin remodeling and transcriptional regulation. This gene has been found to undergo X inactivation. There are four alternatively spliced transcript variants of this gene encoding different isoforms. [provided by RefSeq]
Specifications
Specifications
| Accession Number | AAH31210.1 |
| For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 10943 |
| Molecular Weight (g/mol) | 86.2kDa |
| Name | MSL3L1 (Human) Recombinant Protein (P01) |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | MSASEGMKFKFHSGEKVLCFEPDPTKARVLYDAKIVDVIVGKDEKGRKIPEYLIHFNGWNRSWDRWAAEDHVLRDTDENRRLQRKLARKAVARLRSTGRKKKRCRLPGVDSVLKGLPTEEKDENDENSLSSSSDCSENKDEEISEESDIEEKTEVKEEPELQTRREMEERTITIEIPEVLKKQLEDDCYYINRRKRLVKLPCQTNIITILESYVKHFAINAAFSANERPRHHHVMPHANMNVHYIPAEKNVDLCKEMVDGLRITFDYTLPLVLLYPYEQAQYKKVTSSKFFLPIKESATSTNRSQEELSPSPPLLNPSTPQSTESQPTTGEPATPKRRKAEPEALQSLRRSTRHSANCDRLSESSASPQPKRRQQDTSASMPKLFLHLEKKTPVHSRSSSPIPLTPSKEGSAVFAGFEGRRTNEINEVLSWKLVPDNYPPGDQPPPPSYIYGAQHLLRLFVKLPEILGKMSFSEKNLKALLKHFDLFLRFLAEYHDDFFPESAYVAACEAHYSTKNPRAIY |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.