Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human MT2A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7163325
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10176SP 25 μg
MAB10176100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10176SP Supplier R&D Systems Supplier No. MAB10176SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MT2A Monoclonal specifically detects MT2A in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen MT2A
Applications Immunohistochemistry
Classification Monoclonal
Clone 1006811
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P02795
Gene Alias MT2, MT-2, MT-II
Host Species Mouse
Immunogen Synthetic peptide containing human MT2A, Accession # P02795
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 4502
Test Specificity Detects human MT2A in diect ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.