Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MTERF Full-length ORF (AAH00965, 1 a.a. - 399 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953139 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-139 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-953-139 Supplier Abnova™ Supplier No. H00007978P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a mitochondrial transcription termination factor. This protein participates in attenuating transcription from the mitochondrial genome; this attenuation allows higher levels of expression of 16S ribosomal RNA relative to the tRNA gene downstream. The product of this gene has three leucine zipper motifs bracketed by two basic domains that are all required for DNA binding. There is evidence that, for this protein, the zippers participate in intramolecular interactions that establish the three-dimensional structure required for DNA binding. [provided by RefSeq]

Sequence: MQSLSLGQTSISKGLNYLTIMAPGNLWHMRNNFLFGSRCWMTRFSAENIFKSVSFRLFGVKCHNTDSEPLKNEDLLKNLLTMGVDIDMARKRQPGVFHRMITNEQDLKMFLLSKGASKEVIASIISRYPRAITRTPENLSKRWDLWRKIVTSDLEIVNILERSPESFFRSNNNLNLENNIKFLYSVGLTRKCLCRLLTNAPRTFSNSLDLNKQMVEFLQAAGLSLGHNDPTDFVRKIIFKNPFILIQSTKRVKANIEFLRSTFNLNSEELLVLICGPGAEILDLSNDYARRSYANIKEKLFSLGCTEEEVQKFVLSYPDVIFLAEKKFNDEIDCLMEENISISQIIENPRVLDSSISTLKSRIKELVNAGCNLSTLNITLLSWSKKRYEAKLKKLSRFA

Specifications

Accession Number AAH00965
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7978
Molecular Weight (g/mol) 69.63kDa
Name MTERF (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MQSLSLGQTSISKGLNYLTIMAPGNLWHMRNNFLFGSRCWMTRFSAENIFKSVSFRLFGVKCHNTDSEPLKNEDLLKNLLTMGVDIDMARKRQPGVFHRMITNEQDLKMFLLSKGASKEVIASIISRYPRAITRTPENLSKRWDLWRKIVTSDLEIVNILERSPESFFRSNNNLNLENNIKFLYSVGLTRKCLCRLLTNAPRTFSNSLDLNKQMVEFLQAAGLSLGHNDPTDFVRKIIFKNPFILIQSTKRVKANIEFLRSTFNLNSEELLVLICGPGAEILDLSNDYARRSYANIKEKLFSLGCTEEEVQKFVLSYPDVIFLAEKKFNDEIDCLMEENISISQIIENPRVLDSSISTLKSRIKELVNAGCNLSTLNITLLSWSKKRYEAKLKKLSRFA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC131634
Common Name MTERF
Gene Symbol MTERF
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.