Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MTL5 Full-length ORF (NP_001034745.1, 1 a.a. - 306 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953168 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-168 10 μg
89-953-169 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-168 Supplier Abnova™ Supplier No. H00009633P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Metallothionein proteins are highly conserved low-molecular-weight cysteine-rich proteins that are induced by and bind to heavy metal ions and have no enzymatic activity. They may play a central role in the regulation of cell growth and differentiation and are involved in spermatogenesis. This gene encodes a metallothionein-like protein which has been shown to be expressed differentially in mouse testis and ovary. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MEEGPLPGGLPSPEDAMVTELLSPEGPFASENIGLKAPVKYEEDEFHVFKEAYLGPADPKEPVLHAFNPALGADCKGQVKAKLAGGDSDGGELLGEYPGIPELSALEDVALLQAPQPPACNVHFLSSLLPAHRSPAVLPLGAWVLEGASHPGVRMIPVEIKEAGGTTTSNNPEEATLQNLLAQESCCKFPSSQELEDASCCSLKKDSNPMVICQLKGGTQMLCIDNSRTRELKALHLVPQYQDQNNYLQSDVPKPMTALVGRFLPASTKLNLITQQLEGALPSVVNGSAFPSGSTLPGPPKITLAG

Specifications

Accession Number NP_001034745.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9633
Molecular Weight (g/mol) 58.7kDa
Name MTL5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MEEGPLPGGLPSPEDAMVTELLSPEGPFASENIGLKAPVKYEEDEFHVFKEAYLGPADPKEPVLHAFNPALGADCKGQVKAKLAGGDSDGGELLGEYPGIPELSALEDVALLQAPQPPACNVHFLSSLLPAHRSPAVLPLGAWVLEGASHPGVRMIPVEIKEAGGTTTSNNPEEATLQNLLAQESCCKFPSSQELEDASCCSLKKDSNPMVICQLKGGTQMLCIDNSRTRELKALHLVPQYQDQNNYLQSDVPKPMTALVGRFLPASTKLNLITQQLEGALPSVVNGSAFPSGSTLPGPPKITLAG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CXCDC2/MTLT/TESMIN
Common Name MTL5
Gene Symbol MTL5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.