Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human mu Opioid R/OPRM1 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NB001565 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001565 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001565 Supplier R&D Systems™ Supplier No. NBP278779

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

Specifications

Assay Range 12.50 to 800 pg/mL
Assay Sensitivity 7.50 pg/mL
Product Line Novus
Product Type ELISA Kit (Chemiluminescence)
Sample Type Other Biological Fluids, Plasma, Serum
Specificity This CLIA kit recognizes Human beta-EPR in samples. No significant cross-reactivity or interference between Human beta-EPR and analogues was observed
For Use With (Application) ELISA
Gene ID (Entrez) 4988
Gene Symbol OPRM1
Host Species Human
Quantity 1 Kit
Research Discipline Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Target mu Opioid R/OPRM1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.