Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MUC20 Full-length ORF (AAH44243.1, 1 a.a. - 538 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953237 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-237 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-953-237 Supplier Abnova™ Supplier No. H00200958P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the mucin protein family. Mucins are high molecular weight glycoproteins secreted by many epithelial tissues to form an insoluble mucous barrier. The shorter isoform expressed by this gene is localized to the plasma membrane, whereas the longer isoform might be secreted. The C terminus of this protein associates with the multifunctional docking site of the met proto-oncogene and suppresses activation of some downstream met signaling cascades. The protein features a tandem repeat domain that varies between 2 and 6 copies in most individuals. The allele represented in the human reference assembly contains 12 copies. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MGCLWGLALPLFFFCWEVGVSGSSAGPSTRRADTAMTTDDTEVPAMTLAPGHAALETQTLSAETSSRASTPAGPIPEAETRGAKRISPARETRSFTKTSPNFMVLIATSVETSAASGSPEGAGMTTVQTITGSDPEEAIFDTLCTDDSSEEAKTLTMDILTLAHTSTEAKGLSSESSASSDGPHPVITPSRASESSASSDGPHPVITPSRASESSASSDGPHPVITPSWSPGSDVTLLAEALVTVTNIEVINCSITEIETTTSSIPGASDIDLIPTEGVKASSTSDPPALPDSTEAKPHITEVTASAETLSTAGTTESAAPHATVGTPLPTNSATEREVTAPGATTLSGALVTVSRNPLEETSALSVETPSYVKVSGAAPVSIEAGSAVGKTTSFAGSSASSYSPSEAALKNFTPSETPTMDIATKGPFPTSRDPLPSVPPTTTNSSRGTNSTLAKITTSAKTTMKPPTATPTTARTRPTTDVSAGENGGFLLLRLSVASPEDLTDPRVAERLMQQLHRELHAHAPHFQVSLLRVRRG

Specifications

Accession Number AAH44243.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 200958
Molecular Weight (g/mol) 81.6kDa
Name MUC20 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MGCLWGLALPLFFFCWEVGVSGSSAGPSTRRADTAMTTDDTEVPAMTLAPGHAALETQTLSAETSSRASTPAGPIPEAETRGAKRISPARETRSFTKTSPNFMVLIATSVETSAASGSPEGAGMTTVQTITGSDPEEAIFDTLCTDDSSEEAKTLTMDILTLAHTSTEAKGLSSESSASSDGPHPVITPSRASESSASSDGPHPVITPSRASESSASSDGPHPVITPSWSPGSDVTLLAEALVTVTNIEVINCSITEIETTTSSIPGASDIDLIPTEGVKASSTSDPPALPDSTEAKPHITEVTASAETLSTAGTTESAAPHATVGTPLPTNSATEREVTAPGATTLSGALVTVSRNPLEETSALSVETPSYVKVSGAAPVSIEAGSAVGKTTSFAGSSASSYSPSEAALKNFTPSETPTMDIATKGPFPTSRDPLPSVPPTTTNSSRGTNSTLAKITTSAKTTMKPPTATPTTARTRPTTDVSAGENGGFLLLRLSVASPEDLTDPRVAERLMQQLHRELHAHAPHFQVSLLRVRRG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ14408/KIAA1359
Common Name MUC20
Gene Symbol MUC20
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.