Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MUC5B Partial ORF (XP_039877.9, 4186 a.a. - 4295 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953244 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-244 25 μg
89-953-243 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-244 Supplier Abnova™ Supplier No. H00727897Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Mucins are high molecular mass, highly glycosylated macromolecules that are the major components of mucus secretions. MUC5B is a salivary mucin that is thought to contribute to the lubricating and viscoelastic properties of whole saliva. It is composed of 14.9% protein, 78.1% carbohydrate, and 7% sulfate (Troxler et al., 1995 [PubMed 8554565]).[supplied by OMIM]

Sequence: CEEDSCQVRINTTILWHQGCETEVNITFCEGSCPGASKYSAEAQAMQHQCTCCQERRVHEETVPLHCPNGSAILHTYTHVDECGCTPFCVPAPMAPPHTRGFPAQEATAV

Specifications

Accession Number XP_039877.9
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 727897
Molecular Weight (g/mol) 37.84kDa
Name MUC5B (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen CEEDSCQVRINTTILWHQGCETEVNITFCEGSCPGASKYSAEAQAMQHQCTCCQERRVHEETVPLHCPNGSAILHTYTHVDECGCTPFCVPAPMAPPHTRGFPAQEATAV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MG1/MUC5/MUC9
Common Name MUC5B
Gene Symbol MUC5B
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.