Learn More
American Research Products Inc Human mucosae associated epith
Supplier:  American Research Products Inc CSBE07310H
Encompass
Human mucosae associated epithelia chemokine MEC ELISA Kit from CUSABIO Detection Range 78 pg/ml - 5000 pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29874