Learn More
Sartorius Human Multi Ab Internalization
Catalog No. 502891677 Shop All Sartorius Products
Encompass
No wash protocol- fast easy antibody-labeling method optimal for screening antibody internalization.High throughput evaluation of antibody internalization- Assay designed to measure internalization of a large number of antibodies in 96- and 384-well plate formats using 10 L of cells and 10 L of labeled antibody kit .Simultaneously measure antibody internalization cell specificity and cell health within the same well.PrincipleAntibodies are easily labeled with Intellicyt Antibody Internalization Reagent a novel pH sensitive dye.
Catalog No. 50-289-1677 Supplier Sartorius Supplier No. 97048
No wash protocol- fast easy antibody-labeling method optimal for screening antibody internalization.High throughput evaluation of antibody internalization- Assay designed to measure internalization of a large number of antibodies in 96- and 384-well plate formats using 10 L of cells and 10 L of labeled antibody kit .Simultaneously measure antibody internalization cell specificity and cell health within the same well.PrincipleAntibodies are easily labeled with Intellicyt Antibody Internalization Reagent a novel pH sensitive dye.