Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MXD3 Full-length ORF (NP_112590.1, 1 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953291 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-291 25 μg
89-953-290 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-291 Supplier Abnova™ Supplier No. H00083463P01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the Myc superfamily of basic helix-loop-helix leucine zipper transcriptional regulators. The encoded protein forms a heterodimer with the cofactor MAX which binds specific E-box DNA motifs in the promoters of target genes and regulates their transcription. Disruption of the MAX-MXD3 complex is associated with uncontrolled cell proliferation and tumorigenesis. Transcript variants of this gene encoding different isoforms have been described.[provided by RefSeq]

Sequence: MEPLASNIQVLLQAAEFLERREREAEHGYASLCPHRSPGPIHRRKKRPPQAPGAQDSGRSVHNELEKRRRAQLKRCLERLKQQMPLGADCARYTTLSLLRRARMHIQKLEDQEQRARQLKERLRSKQQSLQRQLEQLRGLAGAAERERLRADSLDSSGLSSERSDSDQEELEVDVESLVFGGEAELLRGFVAGQEHSYSHGGGAWL

Specifications

Accession Number NP_112590.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 83463
Molecular Weight (g/mol) 49.9kDa
Name MXD3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MEPLASNIQVLLQAAEFLERREREAEHGYASLCPHRSPGPIHRRKKRPPQAPGAQDSGRSVHNELEKRRRAQLKRCLERLKQQMPLGADCARYTTLSLLRRARMHIQKLEDQEQRARQLKERLRSKQQSLQRQLEQLRGLAGAAERERLRADSLDSSGLSSERSDSDQEELEVDVESLVFGGEAELLRGFVAGQEHSYSHGGGAWL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BHLHC13/FLJ35523/MAD3/MGC2383/MYX
Common Name MXD3
Gene Symbol MXD3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.