Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MXI1 Full-length ORF (NP_005953.4, 1 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-298 25 ug
89-953-297 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-298 Supplier Abnova™ Supplier No. H00004601P02L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Expression of the c-myc gene, which produces an oncogenic transcription factor, is tightly regulated in normal cells but is frequently deregulated in human cancers. The protein encoded by this gene is a transcriptional repressor thought to negatively regulate MYC function, and is therefore a potential tumor suppressor. This protein inhibits the transcriptional activity of MYC by competing for MAX, another basic helix-loop-helix protein that binds to MYC and is required for its function. Defects in this gene are frequently found in patients with prostate tumors. Three alternatively spliced transcripts encoding different isoforms have been described. Additional alternatively spliced transcripts may exist but the products of these transcripts have not been verified experimentally. [provided by RefSeq]

Sequence: MERVKMINVQRLLEAAEFLERRERECEHGYASSFPSMPSPRLQHSKPPRRLSRAQKHSSGSSNTSTANRSTHNELEKNRRAHLRLCLERLKVLIPLGPDCTRHTTLGLLNKAKAHIKKLEEAERKSQHQLENLEREQRFLKWRLEQLQGPQEMERIRMDSIGSTISSDRSDSEREEIEVDVESTEFSHGEVDNISTTSISDIDDHSSLPSIGSDEGYSSASVKLSFTS

Specifications

Accession Number NP_005953.4
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4601
Molecular Weight (g/mol) 52.5kDa
Name MXI1 (Human) Recombinant Protein (P02)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MERVKMINVQRLLEAAEFLERRERECEHGYASSFPSMPSPRLQHSKPPRRLSRAQKHSSGSSNTSTANRSTHNELEKNRRAHLRLCLERLKVLIPLGPDCTRHTTLGLLNKAKAHIKKLEEAERKSQHQLENLEREQRFLKWRLEQLQGPQEMERIRMDSIGSTISSDRSDSEREEIEVDVESTEFSHGEVDNISTTSISDIDDHSSLPSIGSDEGYSSASVKLSFTS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MAD2/MGC43220/MXD2/MXI/bHLHc11
Common Name MXI1
Gene Symbol MXI1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.