Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MYL5 Full-length ORF (AAH40050.1, 1 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953365 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-365 10 ug
89-953-366 25μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-365 Supplier Abnova™ Supplier No. H00004636P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia. [provided by RefSeq]

Sequence: MDQNRDGFIDKEDLKDTYASLGKTNVKDDELDAMLKEASGPINFTMFLNLFGEKLSGTDAEETILNAFKMLDPDGKGKINKEYIKRLLMSQADKMTAEEVDQMFQFASIDVAGNLDYKALSYVITHGEEKEE

Specifications

Accession Number AAH40050.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4636
Molecular Weight (g/mol) 40.26kDa
Name MYL5 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MDQNRDGFIDKEDLKDTYASLGKTNVKDDELDAMLKEASGPINFTMFLNLFGEKLSGTDAEETILNAFKMLDPDGKGKINKEYIKRLLMSQADKMTAEEVDQMFQFASIDVAGNLDYKALSYVITHGEEKEE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name MYL5
Gene Symbol MYL5
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.