Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MYO5A Partial ORF (NP_000250.1, 1758 a.a. - 1853 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953419 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-419 25 ug
89-953-418 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-419 Supplier Abnova™ Supplier No. H00004644Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene is one of three myosin V heavy-chain genes, belonging to the myosin gene superfamily. Myosin V is a class of actin-based motor proteins involved in cytoplasmic vesicle transport and anchorage, spindle-pole alignment and mRNA translocation. The protein encoded by this gene is abundant in melanocytes and nerve cells. Mutations in this gene cause Griscelli syndrome type-1 (GS1), Griscelli syndrome type-3 (GS3) and neuroectodermal melanolysosomal disease, or Elejalde disease. Multiple alternatively spliced transcript variants encoding different isoforms have been reported, but the full-length nature of some variants has not been determined. [provided by RefSeq]

Sequence: KKTDDDAEAICSMCNALTTAQIVKVLNLYTPVNEFEERVSVSFIRTIQMRLRDRKDSPQLLMDAKHIFPVTFPFNPSSLALETIQIPASLGLGFIS

Specifications

Accession Number NP_000250.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4644
Molecular Weight (g/mol) 36.3kDa
Name MYO5A (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen KKTDDDAEAICSMCNALTTAQIVKVLNLYTPVNEFEERVSVSFIRTIQMRLRDRKDSPQLLMDAKHIFPVTFPFNPSSLALETIQIPASLGLGFIS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias GS1/MYH12/MYO5/MYR12
Common Name MYO5A
Gene Symbol MYO5A
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.