Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Myostatin Propeptide Recombinant Protein

Catalog No. PIRP8653
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8653 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8653 Supplier Invitrogen™ Supplier No. RP8653

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Recombinant Protein

Myostatin Propeptide (human) Recombinant Protein, RP-8653, from Invitrogen.

Myostatin (GDF-8), a member of the TGF-beta superfamily, is a potent and specific negative regulator of skeletal muscle mass. The myostatin propeptide is known to bind and inhibit myostatin in vitro. This interaction is relevant in vivo, with a majority (>70%) of myostatin in serum bound to its propeptide acting as a negative regulator of myostatin. Recombinant human Myostatin Propeptide is a non-glycosylated protein, containing 244 amino acids, with a molecular weight of 27.8 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the ability to inhibit 50 ng/mL of Myostatin on MPC-11 cells and is typically 0.01-0.04 ug/mL. The specific activity of this protein is 2.5x10^5 units/mg. Reconstitute using sterile 20mM HCl at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: MNENSEQKEN VEKEGLCNAC TWRQNTKSSR IEAIKIQILS KLRLETAPNI SKDVIRQLLP KAPPLRELID QYDVQRDDSS DGSLEDDDYH ATTETIITMP TESDFLMQVD GKPKCCFFKF SSKIQYNKVV KAQLWIYLRP VETPTTVFVQ ILRLIKPMKD GTRYTGIRSL KLDMNPGTGI WQSIDVKTVL QNWLKQPESN LGIEIKALDE NGHDLAVTFP GPGEDGLNPF LE.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14793
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.1%TFA with no preservative
Gene ID (Entrez) 2660
Molecular Weight (g/mol) ∽27.8
Name Human Myostatin Propeptide
Quantity 25 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-Myo-Propeptide
Common Name Human Myostatin Propeptide
Gene Symbol MSTN
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.