Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Myostatin Recombinant Protein

Catalog No. PIRP8660
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP8660 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP8660 Supplier Invitrogen™ Supplier No. RP8660

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Recombinant Protein

Myostatin (human) Recombinant Protein, RP-8660, from Invitrogen.

Myostatin is a member of the TGF-beta superfamily that is essential for proper regulation of skeletal muscle mass in a number of species ranging from fish to humans. Myostatin is a secreted protein that negatively regulates skeletal muscle mass determining both muscle fiber number and size. Inhibition of myostatin can increase muscle mass in a number of animal models of human disease, including muscular dystrophy. Recombinant human Myostatin is a non-glycosylated homodimer, containing two 109 amino acid chains, with a total molecular weight of 24.8 kDa. Purity is typically greater than 95% and endotoxin level less than 1 EU/ug. The expected biological activity (ED50) is determined by the inhibition of the proliferation of MPC-11 cells and is typically less than 20 ng/mL. The specific activity of this protein is 5x10^5 units/mg. Reconstitute using sterile 20mM HCl at 0.1mg/mL and centrifuge prior to opening vial. Gently pipet solution down the sides of the vial. DO NOT VORTEX sample. Store reconstituted material at -20°C and add 0.1% BSA for additional stability. Amino acid sequence: DFGLDCDEHS TESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGECEFV FLQKYPHTHL VHQANPRGSA GPCCTPTKMS PINMLYFNGK EQIIYGKIPA MVVDRCGCS.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14793
Concentration 0.1mg/mL
For Use With (Application) Control
Formulation 0.1%TFA with no preservative
Gene ID (Entrez) 2660
Molecular Weight (g/mol) ∽24.8
Name Human Myostatin
Quantity 10 μg
Source E. coli
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias H-Myostatin
Common Name Human Myostatin
Gene Symbol MSTN
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species E. coli
Recombinant Recombinant
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade purified
Show More Show Less

in vitro research only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.