Learn More
Proteintech Group Inc Human N-cadherin ELISA 96well
SDP
Supplier:  Proteintech Group Inc KE0011496T
Encompass_Preferred
Cadherins are a family of transmembrane glycoproteins that mediate calcium-dependent cell-cell adhesion and play an important role in the maintenance of normal tissue architecture. N-cadherin (neural cadherin), also known as CDH2 (cadherin 2), is a 130-kDa transmembrane protein and a classical member of the cadherin superfamily which also include E-, P-, R-, and B-cadherins. Expression of N-cadherin has been reported on various cell types including neurons, endothelial cells and cardiac myocytes. N-cadherin has functions in early brain morphogenesis, synaptogenesis and synaptic plasticity. Soluble N-cadherin (sN-CAD) is found in biological fluids, including serum, seminal fluid samples, and urine. It has been shown that significantly higher amounts of serum sN-CAD are present in cancer patients than in persons with no evidence of disease.
Catalog No. 50-197-4074
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.