Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NAGPA Partial ORF (NP_057340, 309 a.a. - 408 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953558 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-558 10 μg
89-953-559 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-558 Supplier Abnova™ Supplier No. H00051172Q01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Hydrolases are transported to lysosomes after binding to mannose 6-phosphate receptors in the trans-Golgi network. This gene encodes the enzyme that catalyzes the second step in the formation of the mannose 6-phosphate recognition marker on lysosomal hydrolases. Commonly known as 'uncovering enzyme' or UCE, this enzyme removes N-acetyl-D-glucosamine (GlcNAc) residues from GlcNAc-alpha-P-mannose moieties and thereby produces the recognition marker. This reaction most likely occurs in the trans-Golgi network. This enzyme functions as a homotetramer of two disulfide-linked homodimers. In addition to having an N-terminal signal peptide, the protein's C-terminus contains multiple signals for trafficking it between lysosomes, the plasma membrane, and trans-Golgi network. [provided by RefSeq]

Sequence: DNMWRCPRQVSTVVCVHEPRCQPPDCHGHGTCVDGHCQCTGHFWRGPGCDELDCGPSNCSQHGLCTETGCRCDAGWTGSNCSEECPLGWHGPGCQRPCKC

Specifications

Accession Number NP_057340
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51172
Molecular Weight (g/mol) 36.74kDa
Name NAGPA (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen DNMWRCPRQVSTVVCVHEPRCQPPDCHGHGTCVDGHCQCTGHFWRGPGCDELDCGPSNCSQHGLCTETGCRCDAGWTGSNCSEECPLGWHGPGCQRPCKC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias APAA/UCE
Common Name NAGPA
Gene Symbol NAGPA
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.