Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human NAMPT Recombinant Protein, PeproTech®

Catalog No. 130095UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
130-09-5UG 5 μg
13009100UG 100 μg
13-009-01M 2 x 500 μg
13009250UG 250 μg
130-09-25UG 25 μg
13009500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 130-09-5UG Supplier Gibco™ Supplier No. 130095UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

130-09-1MG will be provided as 2 x 500 μg (130-09-500UG). Recombinant human Visfatin is a 52.6 kDa protein containing 466 amino acid residues (isoform 1). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43490
For Use With (Application) Control, Western Blot Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 10135
Molecular Weight (g/mol) 52.6 kDa
Name Human NAMPT
Purification Method Purified
Quantity 5 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias 1110035O14Rik; AI314458; AI480535; DKFZp666B131; EC 2.4.2.12; hypothetical protein; MGC117256; NAmPRTase; NAMPT; nampt1; nampta; NAMPTP1; Nicotinamide phosphoribosyltransferase; nicotinamide phosphoribosyltransferase 1; nicotinamide phosphoribosyltransferase a; nicotinamide phosphoribosyltransferase pseudogene 1; PBEF; Pbef1; pre-B cell-enhancing factor; pre-B-cell colony enhancing factor 1; pre-B-cell colony-enhancing factor 1; Pre-B-cell colony-enhancing factor 1 homolog; pre-B-cell colony-enhancing factor 1 precursor; VF; VISFATIN
Common Name NAMPT
Gene Symbol NAMPT
Biological Activity The ED50 ?was determined by the dose-dependent proliferation of the RPMI 8226 cells. The expected ED50 ?for this effect is 15.0-20.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MPPNTSKVYS YFECREKKTE NSKLRKVKYE ETVFYGLQYI LNKYLKGKVV TKEKIQEAKD VYKEHFQDDV FNEKGWNYIL EKYDGHLPIE IKAVPEGFVI PRGNVLFTVE NTDPECYWLT NWIETILVQS WYPITVATNS REQKKILAKY LLETSGNLDG LEYKLHDFGY RGVSSQETAG IGASAHLVNF KGTDTVAGLA LIKKYYGTKD PVPGYSVPAA EHSTITAWGK DHEKDAFEHI VTQFSSVPVS VVSDSYDIYN ACEKIWGEDL RHLIVSRSTQ APLIIRPDSG NPLDTVLKVL EILGKKFPVT ENSKGYKLLP PYLRVIQGDG VDINTLQEIV EGMKQKMWSI ENIAFGSGGG LLQKLTRDLL NCSFKCSYVV TNGLGINVFK DPVADPNKRS KKGRLSLHRT PAGNFVTLEE GKGDLEEYGQ DLLHTVFKNG KVTKSYSFDE IRKNAQLNIE LEAAHH
Content And Storage -20°C
Activity The ED50 ?was determined by the dose-dependent proliferation of the RPMI 8226 cells. The expected ED50 ?for this effect is 15.0-20.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.