Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NAP1L4 Partial ORF (NP_005960, 1 a.a. - 82 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953606 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-606 25 ug
89-953-605 10 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-606 Supplier Abnova™ Supplier No. H00004676Q01L

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the nucleosome assembly protein (NAP) family which can interact with both core and linker histones. It can shuttle between the cytoplasm and nucleus, suggesting a role as a histone chaperone. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. [provided by RefSeq]

Sequence: MADHSFSDGVPSDSVEAAKNASNTEKLTDQVMQNPRVLAALQERLDNVPHTPSSYIETLPKAVKRRINALKQLQVRCAHIEA

Specifications

Accession Number NP_005960
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 4676
Molecular Weight (g/mol) 34.76kDa
Name NAP1L4 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen MADHSFSDGVPSDSVEAAKNASNTEKLTDQVMQNPRVLAALQERLDNVPHTPSSYIETLPKAVKRRINALKQLQVRCAHIEA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC4565/NAP2/NAP2L/hNAP2
Common Name NAP1L4
Gene Symbol NAP1L4
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.