Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NBEA Partial ORF (NP_056493.3, 1133 a.a. - 1220 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953661 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-661 25 μg
89-953-660 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-661 Supplier Abnova™ Supplier No. H00026960Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of a large, diverse group of A-kinase anchor proteins that target the activity of protein kinase A to specific subcellular sites by binding to its type II regulatory subunits. Brain-specific expression and coat protein-like membrane recruitment of a highly similar protein in mouse suggest an involvement in neuronal post-Golgi membrane traffic. Mutations in this gene may be associated with a form of autism. Alternatively spliced transcript variants may exist, but their full-length nature has not been determined. [provided by RefSeq]

Sequence: ADEKEDLPNSSTSFLFDKIPKQEEKLLPELSSNHIIPNIQDTQVHLGVSDDLGLLAHMTGSVDLTCTSSIIEEKEFKIHTTSDGMSSI

Specifications

Accession Number NP_056493.3
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26960
Molecular Weight (g/mol) 35.42kDa
Name NBEA (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen ADEKEDLPNSSTSFLFDKIPKQEEKLLPELSSNHIIPNIQDTQVHLGVSDDLGLLAHMTGSVDLTCTSSIIEEKEFKIHTTSDGMSSI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BCL8B/LYST2
Common Name NBEA
Gene Symbol NBEA
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.