Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NCALD Full-length ORF (NP_001035714.1, 1 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89953673 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-953-673 10 μg
89-953-674 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-953-673 Supplier Abnova™ Supplier No. H00083988P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the neuronal calcium sensor (NCS) family of calcium-binding proteins. The protein contains an N-terminal myristoylation signal and four EF-hand calcium binding loops. The protein is cytosolic at resting calcium levels; however, elevated intracellular calcium levels induce a conformational change that exposes the myristoyl group, resulting in protein association with membranes and partial co-localization with the perinuclear trans-golgi network. The protein is thought to be a regulator of G protein-coupled receptor signal transduction. Several alternatively spliced variants of this gene have been determined, all of which encode the same protein; additional variants may exist but their biological validity has not been determined. [provided by RefSeq]

Sequence: MGKQNSKLRPEVMQDLLESTDFTEHEIQEWYKGFLRDCPSGHLSMEEFKKIYGNFFPYGDASKFAEHVFRTFDANGDGTIDFREFIIALSVTSRGKLEQKLKWAFSMYDLDGNGYISKAEMLEIVQAIYKMVSSVMKMPEDESTPEKRTEKIFRQMDTNRDGKLSLEEFIRGAKSDPSIVRLLQCDPSSAGQF

Specifications

Accession Number NP_001035714.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 83988
Molecular Weight (g/mol) 48.6kDa
Name NCALD (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MGKQNSKLRPEVMQDLLESTDFTEHEIQEWYKGFLRDCPSGHLSMEEFKKIYGNFFPYGDASKFAEHVFRTFDANGDGTIDFREFIIALSVTSRGKLEQKLKWAFSMYDLDGNGYISKAEMLEIVQAIYKMVSSVMKMPEDESTPEKRTEKIFRQMDTNRDGKLSLEEFIRGAKSDPSIVRLLQCDPSSAGQF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC33870/MGC74858
Common Name NCALD
Gene Symbol NCALD
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.