Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human NCAM-1/CD56 (NP_000606) VersaClone cDNA
SDP

Catalog No. RDC2495 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDC2495 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RDC2495 Supplier R&D Systems™ Supplier No. RDC2495

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Human NCAM-1/CD56 (NP_000606) VersaClone cDNA clone contains the complete ORF for NCAM-1/CD56, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The NCAM-1/CD56 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided

Specifications

Gene hNCAM-1/CD56
Accession Number NP_000606
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 4684
Content And Storage Store the unopened product at -20 to -80°C
Species Human
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.